Sodium diethyldithiocarbamate trihydrate ACS

Warning

Synonyms: (Diethyldithiocarbamato-S,S')sodium trihydrate, Diethyldithiocarbamic acid sodium salt trihydrate, N,N-Diethyldithiocarbamic acid sodium salt trihydrate

228680-100G 228680-500G 228680-5G
220045-791EA 90 USD
220045-791 220045-792 220045-793
Sodium diethyldithiocarbamate trihydrate ACS
Sodium diethyldithiocarbamate trihydrate
Formula: (C₂H₅)₂NCSSNa·3H₂O
MW: 225.31 g/mol
Melting Pt: 95 - 98.5 °C
Density: 1.1 g/cm³ (20 °C)
MDL Number: MFCD00150617
CAS Number: 20624-25-3
EINECS: 205-710-6
UN: 3077
ADR: 9,III
Merck Index: 12,03443

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR