Magnesium acetate tetrahydrate ≥98% ACS

228648-500G 228648-2.5KG
220045-786EA 102.15 USD
220045-786 220045-784 220045-785
Magnesium acetate tetrahydrate ≥98% ACS
Magnesium acetate tetrahydrate
Formula: (H₃CCOO)₂Mg·4H₂O
Melting Pt: 80 °C
Density: 1.454 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149214
CAS Number: 16674-78-5
EINECS: 205-554-9
Merck Index: 13,05676

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR