3-Chloro-4-fluoroaniline ≥98%

Warning

228583-100G 228583-25G
220045-778EA 66.45 USD
220045-778 220045-779
3-Chloro-4-fluoroaniline ≥98%
3-Chloro-4-fluoroaniline
Formula: C₆H₅ClFN
MW: 145.56 g/mol
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 41 - 44 °C
Density: 1.226 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007767
CAS Number: 367-21-5
EINECS: 206-682-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR