5-Bromonicotinic acid

Synonyms: 3-Bromo-5-pyridine carboxylic acid, 5-Bromopyridine-3-carboxylic acid, 5-Bromo-3-pyridinecarboxylic acid

228435-10G 228435-50G
220045-767EA 70.22 USD
220045-767 220045-768
5-Bromonicotinic acid
5-Bromonicotinic acid
Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 184…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00009783
CAS Number: 20826-04-4
EINECS: 244-065-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR