Chloroacetamide ≥98%

Danger

Synonyms: 2-Chloroacetamide

22790-250G-F 22790-U
220045-036EA 36.18 USD
220045-036 220045-037
Chloroacetamide ≥98%
Chloroacetamide
Formula: C₂H₄ClNO
MW: 93.51 g/mol
Boiling Pt: 256 °C (1013 hPa)
Melting Pt: 115 - 117 °C
Density: 1.6 g/cm³ (20 °C)
Flash Pt: 170 °C
Storage Temperature: Ambient
MDL Number: MFCD00008027
CAS Number: 79-07-2
EINECS: 201-174-2
UN: 2811
ADR: 6.1,III
Merck Index: 13,02127

Order Now

Specification Test Results

Assay 98% HPLC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR