Silver chromate

227897-10G 227897-50G
220045-300EA 122.92 USD
220045-300 220045-301
Silver chromate
Silver chromate
Formula: Ag₂CrO₄
Boiling Pt: 1550 °C (1013 hPa)
Density: 5.536 g/cm³ (25 °C)
MDL Number: MFCD00003402
CAS Number: 7784-01-2
EINECS: 232-043-8
UN: 1479
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR