Nickel (II) sulfate hexahydrate ACS

Danger

227676-2.5KG 227676-500G
220045-284EA 356.92 USD
220045-284 220045-285 220045-283
Nickel (II) sulfate hexahydrate ACS
Nickel(II) sulphate hexahydrate
Formula: NiSO₄·6H₂O
MW: 262.85 g/mol
Melting Pt: 840 °C
Density: 2.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149813
CAS Number: 10101-97-0
EINECS: 232-104-9
UN: 3288
ADR: 6.1,III
Merck Index: 13,06541

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR