Aluminium sulfate octadecahydrate ≥98% ACS

Danger

Synonyms: Trade termforalum, Aluminum(III) sulphate octadecahydrate, Aluminum sulphate octadecahydrate

227617-2.5KG 227617-500G 227617-100G
220045-280EA 492.92 USD
220045-280 220045-281 220045-279
Aluminium sulfate octadecahydrate ≥98% ACS
Aluminium sulphate octadecahydrate
Formula: Al₂(SO₄)₃·18H₂O
MW: 666.43 g/mol
Melting Pt: 86 °C
Density: 1.69 g/cm³ (20 °C)
MDL Number: MFCD00149136
CAS Number: 7784-31-8
EINECS: 233-135-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4dace68759944aa1c6c0f56a372ea396f004e261&vid=e63de3c58650a97622a605c237aa52402ff157e4&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR