4-Bromobutyl acetate ≥98%

Synonyms: Acetic acid 4-bromobutyl ester

227579-10G 227579-50G
220045-276EA 85.08 USD
220045-276 220045-277
4-Bromobutyl acetate ≥98%
4-Bromobutyl acetate
Formula: C₆H₁₁BrO₂
MW: 195.06 g/mol
MDL Number: MFCD00000263
CAS Number: 4753-59-7
EINECS: 225-277-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR