2,6-Bis(hydroxymethyl)-p-cresol ≥95%

Synonyms: 4-Methyl-2,6-bis(hydroxymethyl)phenol, 1,3-Bis(hydroxymethyl)-2-hydroxy-5-methylbenzene, 2,6-Bis(hydroxymethyl)-4-methylphenol, 3,5-Bis(hydroxymethyl)-4-hydroxytoluene

227528-25G 227528-100G 227528-250G
220045-034EA 70.97 USD
220045-034 220045-032 220045-033
2,6-Bis(hydroxymethyl)-p-cresol ≥95%
2,6-Bis(hydroxymethyl)-p-cresol
Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 363 °C (1013 hPa)
Melting Pt: 133…134 °C
Density: 1.382 g/cm³ (-173 °C)
MDL Number: MFCD00004619
CAS Number: 91-04-3
EINECS: 202-036-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR