Silver benzoate ≥99%

Synonyms: Benzoic acid silver (I) salt

227277-50G 227277-10G
220045-690EA 306.92 USD
220045-690 220045-689
Silver benzoate ≥99%
Silver benzoate
Formula: C₇H₅AgO₂
MW: 228.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013030
CAS Number: 532-31-0
EINECS: 208-533-2
Merck Index: 13,01092

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR