Benzyl isocyanate ≥99%

227269-25G 227269-5G
220045-687EA 215.38 USD
220045-687 220045-688
Benzyl isocyanate ≥99%
Benzyl isocyanate
Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 101 °C (1013 hPa)
Density: 1.0838 g/cm³ (20 °C)
Flash Pt: 45 °C
MDL Number: MFCD00009701
CAS Number: 3173-56-6
EINECS: 221-640-9
UN: 3080
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR