Silver nitrite

227188-10G 227188-50G
220045-672EA 109.23 USD
220045-672 220045-673
Silver nitrite
Silver nitrite
Formula: AgNO₂
MW: 153.87 g/mol
Melting Pt: 140 °C
Density: 4.45 g/cm³ (20 °C)
MDL Number: MFCD00003413
CAS Number: 7783-99-5
EINECS: 232-041-7
UN: 2627
ADR: 5.1,II
Merck Index: 12,08663

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR