Vanadium(III) acetylacetonate

Synonyms: Vanadium(III) acetylacetonate, Vanadium (III) tris(2,4-pentanedionato-O,O'), Vanadium (III) 2,4-pentanedionate, Tris(2,4-pentanedionato-O,O')vanadium (III)

227110-50G 227110-10G
220045-664EA 212.31 USD
220045-664 220045-663
Vanadium(III) acetylacetonate
Vanadium(III) acetylacetonate
Formula: C₁₅H₂₁O₆V
MW: 348.27 g/mol
Boiling Pt: 170 °C (0.05 mmHg)
Melting Pt: 185…190 °C
Density: ~ 1 g/cm³ (20 °C)
MDL Number: MFCD00000033
CAS Number: 13476-99-8
EINECS: 236-759-1
UN: 3285
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR