Quinoxaline

22710-U 22710-100G-F
220045-660EA 42 USD
220045-660 220045-659
Quinoxaline
Quinoxaline
Formula: C₈H₆N₂
MW: 130.15 g/mol
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 30…32 °C
Density: 1.1485 g/cm³ (32 °C)
MDL Number: MFCD00006719
CAS Number: 91-19-0
EINECS: 202-047-4
Merck Index: 12,08263

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR