3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS) hydrate

226947-1G 226947-5G
220045-589EA 73.02 USD
220045-589 220044-391
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS) hydrate
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS) hydrate
Formula: C₃₂H₅₈N₂O₇S·H₂O
MW: 632.9 g/mol
Storage Temperature: Refrigerator
CAS Number: 313223-04-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR