Silver fluoride

Warning

Synonyms: Silver monofluoride, Silver(I) fluoride, Argentous fluoride

226866-5G 226866-25G
220045-578EA 156.92 USD
220045-578 220045-577
Silver fluoride
Silver fluoride
Formula: AgF
MW: 126.87 g/mol
Boiling Pt: 1150 °C (1013 hPa)
Melting Pt: 433 °C
Density: 5.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003410
CAS Number: 7775-41-9
EINECS: 231-895-8
UN: 3260
ADR: 8,II
Merck Index: 12,08657

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR