Phosphorus(V) bromide ≥95%

226734-100G 226734-25G
220045-559EA 311.23 USD
220045-559 220045-560
Phosphorus(V) bromide ≥95%
Phosphorus(V) bromide
Formula: PBr₅
Melting Pt: ? 100 °C
Density: 3.61 g/cm³ (20 °C)
MDL Number: MFCD00011437
CAS Number: 7789-69-7
EINECS: 232-186-6
UN: 2691
ADR: 8,II
Merck Index: 12,07507

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR