Butyl nitrite ≥95%

Danger

226637-100G 226637-25G
220045-551EA 202.46 USD
220045-551 220045-552
Butyl nitrite ≥95%
Butyl nitrite
Formula: C₄H₉NO₂
Boiling Pt: 75 °C (1013 hPa)
Density: 0.88 g/cm³ (20 °C)
Flash Pt: 10 °C
MDL Number: MFCD00002058
CAS Number: 544-16-1
EINECS: 208-862-1
UN: 2351
ADR: 3,II
Merck Index: 12,01617

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR