(±)-1-Phenyl-2-propyn-1-ol ≥98%

Synonyms: 1-Phenyl-2-propyn-1-ol

226610-10G 226610-1G
220045-549EA 186.31 USD
220045-549 220045-550
(±)-1-Phenyl-2-propyn-1-ol ≥98%
(±)-1-Phenyl-2-propyn-1-ol
Formula: C₉H₈O
MW: 132.16 g/mol
MDL Number: MFCD00021860
CAS Number: 4187-87-5
EINECS: 224-064-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR