(Chloromethyl)dimethylchlorosilane

Synonyms: CMDMCS, Chloro(chloromethyl)dimethylsilane

226181-100G 226181-25G
220045-254EA 236.92 USD
220045-254 220045-255
(Chloromethyl)dimethylchlorosilane
(Chloromethyl)dimethylchlorosilane
Formula: C₃H₈Cl₂Si
MW: 143.09 g/mol
MDL Number: MFCD00000875
CAS Number: 1719-57-9
EINECS: 217-006-6
UN: 2985
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR