Trimethylsilyl trifluoromethanesulfonate ≥99%

Synonyms: Trimethylsilyl trifluoromethanesulphonate

225649-10G 225649-50G
220045-485EA 46.31 USD
220045-485 220045-486
Trimethylsilyl trifluoromethanesulfonate ≥99%
Trimethylsilyl trifluoromethanesulphonate
Formula: C₄H₉F₃O₃SSi
MW: 222.26 g/mol
Boiling Pt: 77 °C (1013 hPa)
Density: 1.225 g/cm³ (20 °C)
MDL Number: MFCD00000406
CAS Number: 27607-77-8
EINECS: 248-565-4
Merck Index: 12,09849

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR