Quinaldine

Warning

Synonyms: 2-Methylquinoline

22550-250ML-F 22550-U 22550-1L-F
220045-470EA 156.15 USD
220045-470 220045-471 220045-469
Quinaldine
Quinaldine
Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 247.6 °C (1013 hPa)
Melting Pt: –2 °C
Density: 1.0585 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00006756
CAS Number: 91-63-4
EINECS: 202-085-1
Merck Index: 12,08228

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR