Biphenyl-4,4'-dicarboxylic acid ≥97%

Synonyms: 4,4'-Biphenyldicarboxylic acid, [1,1'-Biphenyl]-4,4'-dicarboxylic acid

225266-5G 225266-25G
220045-453EA 145.15 USD
220045-453 220045-452
Biphenyl-4,4'-dicarboxylic acid ≥97%
Biphenyl-4,4'-dicarboxylic acid
Formula: C₁₄H₁₀O₄
MW: 242.23 g/mol
MDL Number: MFCD00002554
CAS Number: 787-70-2
EINECS: 212-328-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR