6-Chloro-m-toluidine ≥99%

225118-25G 225118-5G
220045-369EA 924 USD
220045-369 220045-370
6-Chloro-m-toluidine ≥99%
6-Chloro-m-toluidine
Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 228…230 °C (1013 hPa)
Melting Pt: 30 °C
MDL Number: MFCD00007674
CAS Number: 95-81-8
EINECS: 202-454-7
UN: 2239
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc4f3e7d19195ef2f0c03e1a9bd73e315a1480ae&vid=9fbb7000f8a3094cd4d37543991d289a70579707&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR