Iodine monobromide

Danger

224847-250G 224847-50G
220044-879EA 318.77 USD
220044-879 220044-880
Iodine monobromide
Iodine monobromide
Formula: IBr
MW: 206.81 g/mol
Boiling Pt: 116 °C (1013 hPa)
Melting Pt: 37 - 40 °C
Density: 4.41 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011353
CAS Number: 7789-33-5
EINECS: 232-159-9
UN: 3260
ADR: 8,II
Merck Index: 13,05038

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR