Bismuth(III) chloride ≥98%, Reagent Grade

Warning

Synonyms: Bismuth(III) chloride, Bismuth trichloride

224839-100G 224839-25G 224839-500G
220044-389EA 201.23 USD
220044-389 220044-390 220044-877
Bismuth(III) chloride ≥98%, Reagent Grade
Bismuth(III) chloride
Formula: BiCl₃
MW: 315.34 g/mol
Boiling Pt: 447 °C (1013 hPa)
Melting Pt: 232 °C
Density: 4.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003461
CAS Number: 7787-60-2
EINECS: 232-123-2
UN: 3260
ADR: 8,III
Merck Index: 13,01261

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR