Ammonium hydrogen difluoride, Reagent Grade

Danger

224820-2.5KG 224820-500G
220044-386EA 23.04 USD
220044-386 220044-385 220044-387
Ammonium hydrogen difluoride, Reagent Grade
Ammonium hydrogen difluoride
Formula: NH₄HF₂
Melting Pt: 127 °C
Density: 0.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012018
CAS Number: 1341-49-7
EINECS: 215-676-4
UN: 1727
ADR: 8,II
Merck Index: 13,00498

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR