5-Chlorouracil

Synonyms: 5-Chloropyrimidine-2,4(1H,3H)-dione, Chlorouracil

224588-25G 224588-5G
220045-348EA 179.85 USD
220045-348 220045-349
5-Chlorouracil
5-Chlorouracil
Formula: C₄H₃ClN₂O₂
MW: 146.53 g/mol
Melting Pt: ? 300 °C
MDL Number: MFCD00006019
CAS Number: 1820-81-1
EINECS: 217-339-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR