(4-Hydroxyphenyl)acetic acid methyl ester ≥99%

Synonyms: Methyl 4-hydroxyphenylacetate, Methyl (4-hydroxyphenyl)acetate, Methyl 2-(4-hydroxyphenyl)acetate

224502-100G 224502-25G
220045-343EA 177.69 USD
220045-343 220045-344
(4-Hydroxyphenyl)acetic acid methyl ester ≥99%
(4-Hydroxyphenyl)acetic acid methyl ester
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Number: MFCD00002387
CAS Number: 14199-15-6
EINECS: 238-050-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR