D-(+)-Menthol ≥99%

Synonyms: (1S)-(+)-Menthol, (1R,2S,4S)-(+)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, (1S,2R,5S)-(+)-Menthol, D(+)-Menthyl alcoholol

224464-10G 224464-50G
220043-749EA 75.49 USD
220043-749 220043-750
D-(+)-Menthol ≥99%
D-(+)-Menthol
Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00062983
CAS Number: 15356-60-2
EINECS: 239-387-8

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR