Zirconyl chloride octahydrate 98%, Reagent Grade, Sigma-Aldrich®

Danger

Synonyms: Zirconium(IV) oxide chloride octahydrate, Zirconium dichloride oxide octahydrate, Zirconium(IV) oxychloride octahydrate

224316-100G 224316-5G 224316-500G
220045-223EA 91.08 USD
220045-223 220045-225 220045-224
Zirconyl chloride octahydrate 98%, Reagent Grade, Sigma-Aldrich®
Zirconyl chloride octahydrate
Formula: Cl₂OZr·8H₂O
MW: 322.25 g/mol
Melting Pt: > 150 °C
Density: 1.91 g/cm³ (20 °C)
MDL Number: MFCD00149898
CAS Number: 13520-92-8
EINECS: 231-717-9
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR