2,6-Toluenediisocyanate ≥97%

Synonyms: 2,6-TDI, 2-Methyl-m-phenylene diisocyanate, Tolylene 2,6-diisocyanate

223867-1G 223867-5G
220045-187EA 81.2 USD
220045-187 220045-188
2,6-Toluenediisocyanate ≥97%
2,6-Toluenediisocyanate
Formula: C₉H₆N₂O₂
MW: 174.16 g/mol
MDL Number: MFCD00002010
CAS Number: 91-08-7
EINECS: 202-039-0
UN: 2078
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR