Lithium iodide hydrate ≥98%

223816-100G 223816-500G
220045-181EA 83.77 USD
220045-181 220045-182
Lithium iodide hydrate ≥98%
Lithium iodide hydrate
Formula: ILi·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00149769
CAS Number: 17023-25-5
EINECS: 233-822-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR