Ammonium tetrafluoroborate

Synonyms: Ammonium fluoroborate, ammonium fluoborate ammonium borofluoride ammonium fluoroborate [qr] ammonium tetrafluoroborate

223727-100G 223727-500G
220045-170EA 52.77 USD
220045-170 220045-171
Ammonium tetrafluoroborate
Ammonium tetrafluoroborate
Formula: H₄BF₄N
MW: 104.84 g/mol
Melting Pt: 510 °C
Density: 1.87 g/cm³ (20 °C)
MDL Number: MFCD06245785
CAS Number: 13826-83-0
EINECS: 237-531-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR