Silver(I) oxide

Danger

Synonyms: di-Silver oxide, Disilver oxide, Silver oxide

223638-10G 223638-50G
220045-154EA 193.85 USD
220045-154 220045-155
Silver(I) oxide
Silver(I) oxide
Formula: Ag₂O
MW: 231.74 g/mol
Melting Pt: 800…830 °C
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00003404
CAS Number: 20667-12-3
EINECS: 243-957-1
UN: 1479
ADR: 5.1,II
Merck Index: 12,08665

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR