Calcium chloride dihydrate ≥99% ACS

Warning

223506-500G 223506-6X500G 223506-2.5KG
220044-867EA 20.76 USD
220044-867 220044-868 220044-869 220044-866
Calcium chloride dihydrate ≥99% ACS
Calcium chloride dihydrate
Formula: CaCl₂·2H₂O
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 176 °C
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR