di-Potassium oxalate monohydrate ≥99% ACS

Warning

Synonyms: Oxalic acid potassium salt monohydrate, Potassium oxalate hydrate, Potassium oxalate monohydrate, Potassium ethanedioate monohydrate

223425-2.5KG 223425-500G
220045-151EA 241.23 USD
220045-151 220045-152
di-Potassium oxalate monohydrate ≥99% ACS
di-Potassium oxalate monohydrate
Formula: (COOK)₂·H₂O
MW: 184.23 g/mol
Melting Pt: 356 °C
Density: 2.154 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150033
CAS Number: 6487-48-5
EINECS: 209-506-8
UN: 2811
ADR: 6.1,III
Merck Index: 12,07820

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR