γ-Terpinene

Danger

Synonyms: gamma-Terpinene, p-Mentha-1,4-diene

223190-5ML 223190-100ML
220045-127EA 29.83 USD
220045-127 220045-126
γ-Terpinene
γ-Terpinene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 182 °C (1013 hPa)
Density: 0.846 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001537
CAS Number: 99-85-4
EINECS: 202-794-6
UN: 2319
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR