4-Fluoro-N-methylaniline ≥97%

Synonyms: N-(4-Fluorophenyl)-N-methylamine

223069-1G 223069-5G
220045-115EA 60.63 USD
220045-115 220045-116
4-Fluoro-N-methylaniline ≥97%
4-Fluoro-N-methylaniline
Formula: C₇H₈FN
MW: 125.15 g/mol
MDL Number: MFCD00017942
CAS Number: 459-59-6
EINECS: 207-294-1
UN: 2511
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR