2,4,6-Tri-tert-butylaniline ≥99%

Warning

Synonyms: 2,4,6-Tris(tert-butyl)aniline

223050-1G 223050-5G
220045-113EA 45.23 USD
220045-113 220045-114
2,4,6-Tri-tert-butylaniline ≥99%
2,4,6-Tri-tert-butylaniline
Formula: C₁₈H₃₁N
MW: 261.45 g/mol
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 145 °C
Density: 0.991 g/cm³ (19.85 °C)
MDL Number: MFCD00011645
CAS Number: 961-38-6
EINECS: 213-507-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR