1-Phenyl-1,2-propanedione ≥99%

Warning

Synonyms: 1-Phenylpropane-1,2-dione

223034-5G 223034-1G
220044-122EA 113.08 USD
220044-122 220044-121
1-Phenyl-1,2-propanedione ≥99%
1-Phenyl-1,2-propanedione
Formula: C₆H₅C(O)C(O)CH₃
MW: 148.16 g/mol
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: ? 20 °C
Density: 1.116 g/cm³ (20 °C)
MDL Number: MFCD00008755
CAS Number: 579-07-7
EINECS: 209-435-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1394d5392b739ebd2496078378d299949ea575ae&vid=b85d2a588e386bc41f5caca908cd1530a3da9131&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR