4-(Methylthio)benzaldehyde ≥95%

222771-10G 222771-50G
220044-177EA 47.71 USD
220044-177 220044-178
4-(Methylthio)benzaldehyde ≥95%
4-(Methylthio)benzaldehyde
Formula: C₈H₈OS
MW: 152.22 g/mol
Boiling Pt: 267 °C (1013 hPa)
Density: 1.14 g/cm³ (20 °C)
MDL Number: MFCD00006948
CAS Number: 3446-89-7
EINECS: 222-365-7
UN: 3334
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR