3-Fluoro-p-toluidine ≥99%

Synonyms: 3-Fluoro-4-methylaniline, 4-Amino-2-fluorotoluene

222666-25G 222666-5G
220044-165EA 75.46 USD
220044-165 220044-166
3-Fluoro-p-toluidine ≥99%
3-Fluoro-p-toluidine
Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 97 °C (15 mmHg)
Melting Pt: 37 °C
Density: 1.09 g/cm³ (20 °C)
MDL Number: MFCD00007762
CAS Number: 452-77-7
EINECS: 207-212-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR