trans-2-Aminocyclohexanol hydrochloride ≥99%

222577-25G 222577-5G
220044-162EA 236.92 USD
220044-162 220044-163
trans-2-Aminocyclohexanol hydrochloride ≥99%
trans-2-Aminocyclohexanol hydrochloride
Formula: C₆H₁₄ClNO
MW: 151.64 g/mol
Melting Pt: 174…174 °C
MDL Number: MFCD00001490
CAS Number: 5456-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR