3-Methylxanthine

Warning

Synonyms: 3,7-Dihydro-3-methyl-1H-purine-2,6-dione, 2,6-Dihydroxy-3-methylpurine

222526-250MG 222526-1G
220044-380EA 117.38 USD
220044-380 220044-379
3-Methylxanthine
3-Methylxanthine
Formula: C₆H₆N₄O₂
MW: 166.14 g/mol
MDL Number: MFCD00005580
CAS Number: 1076-22-8
EINECS: 214-058-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR