N-(1-Naphthyl)ethylenediamine dihydrochloride ACS

Warning

222488-25G 222488-5G 222488-100G 222488-10G
220044-372EA 124.62 USD
220044-372 220044-373 220044-370 220044-371
N-(1-Naphthyl)ethylenediamine dihydrochloride ACS
N-(1-Naphthyl)ethylenediamine dihydrochloride
Formula: C₁₂H₁₆Cl₂N₂
Melting Pt: 188…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00012556
CAS Number: 1465-25-4
EINECS: 215-981-2
Merck Index: 12,06496
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3643e6df3e9cf8dc627823e9090c97004ee25308&vid=a30fc28279c11af12c6b735b00b84fc8f08caf34&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR