Trisodium phosphate dodecahydrate ≥98% ACS

Warning

Synonyms: Sodium Phosphate tribasic 12-Hydrate, Sodium phosphate dodecahydrate, Sodium phosphate tribasic dodecahydrate

222003-2.5KG 222003-500G
220044-945EA 272.62 USD
220044-945 220044-946
Trisodium phosphate dodecahydrate ≥98% ACS
Trisodium phosphate dodecahydrate
Formula: Na₃PO₄·12H₂O
MW: 380.12 g/mol
Melting Pt: ∼75 °C
Density: 1.62 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149198
CAS Number: 10101-89-0
EINECS: 231-509-8
ADR: 8,III
REACH: 01-2119489800-32-xxxx
Merck Index: 13,08736

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR