Zirconium tetrachloride

Danger

Synonyms: Zirconium (IV) chloride

221880-5G 221880-100G 221880-500G
220043-376EA 83.69 USD
220043-376 220043-832 220043-375
Zirconium tetrachloride
Zirconium tetrachloride
Formula: Cl₄Zr
MW: 233.03 g/mol
Boiling Pt: 3577 °C (1013 hPa)
Melting Pt: 437 °C
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011306
CAS Number: 10026-11-6
EINECS: 233-058-2
UN: 2503
ADR: 8,III
Merck Index: 12,10307

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR