Potassium fluoride dihydrate ≥98%, Reagent Grade

Danger

221872-500G 221872-100G
220043-830EA 118.15 USD
220043-830 220043-829
Potassium fluoride dihydrate ≥98%, Reagent Grade
Potassium fluoride dihydrate
Formula: FK·2H₂O
MW: 94.13 g/mol
Boiling Pt: 1505 °C (1013 hPa)
Melting Pt: 846 °C
Density: 2.45 g/cm³ (20 °C)
MDL Number: MFCD00149929
CAS Number: 13455-21-5
EINECS: 232-151-5
UN: 1812
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR